2-Bromo-3,5-dimethylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3,5-lutidine

A112109-10G A112109-1G A112109-25G
77025-804EA 67.9 USD
77025-804 77025-806 77025-808
2-Bromo-3,5-dimethylpyridine 97%
2-Bromo-3,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.049 g/mol
MDL Number: MFCD00234308
CAS Number: 92992-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR