1-(5-Bromopyrimidin-2-yl)propan-1-one 95%

Supplier: Ambeed
Warning

A111908-100MG A111908-1G A111908-250MG
76982-770EA 57.29 USD
76982-770 76982-772 76982-774
1-(5-Bromopyrimidin-2-yl)propan-1-one 95%
1-(5-Bromopyrimidin-2-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
MDL Number: MFCD19688634
CAS Number: 1346808-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR