3-Methylquinolin-8-ol 95%

Supplier: Ambeed
Warning

A111876-100MG A111876-250MG
77008-950EA 65 USD
77008-950 77008-952
3-Methylquinolin-8-ol 95%
3-Methylquinolin-8-ol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD05662429
CAS Number: 75457-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR