Potassium (1-methyl-4-trifluoroboratomethyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro((4-methylpiperazin-1-yl)methyl)borate

A111830-1G A111830-5G
77025-792EA 33.95 USD
77025-792 77025-794
Potassium (1-methyl-4-trifluoroboratomethyl)piperazine 98%
Potassium (1-methyl-4-trifluoroboratomethyl)piperazine

Precaución: for Research Use Only

Formula: C₆H₁₃BF₃KN₂
MW: 220.0853 g/mol
MDL Number: MFCD10700160
CAS Number: 1015484-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR