2-Iodo-5-trifluoromethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-5-(trifluoromethyl)pyridine

A111804-1G A111804-250MG A111804-25G A111804-5G
76982-762EA 11.67 USD
76982-762 76982-764 76982-766 76982-768
2-Iodo-5-trifluoromethylpyridine 98%
2-Iodo-5-trifluoromethylpyridine

Precaución: for Research Use Only

Formula: C₆H₃F₃IN
MW: 272.99 g/mol
MDL Number: MFCD08741351
CAS Number: 100366-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR