2-Chloro-6-(difluoromethoxy)pyridine 97%

Supplier: Ambeed
Warning

A111616-100MG A111616-1G A111616-250MG
76982-746EA 82.76 USD
76982-746 76982-748 76982-750
2-Chloro-6-(difluoromethoxy)pyridine 97%
2-Chloro-6-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₂NO
MW: 179.552 g/mol
MDL Number: MFCD13185850
CAS Number: 1214377-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR