2-Fluoro-3-iodophenylboronic acid 97%

Supplier: Ambeed

Synonyms: (2-Fluoro-3-iodophenyl)boronic acid

A111502-1G A111502-25G A111502-5G
76995-246EA 12.74 USD
76995-246 76995-248 76995-250
2-Fluoro-3-iodophenylboronic acid 97%
2-Fluoro-3-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BFIO₂
MW: 265.82 g/mol
MDL Number: MFCD05664305
CAS Number: 1016231-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR