tert-Butyl 7-fluoro-6-nitro-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%

Supplier: Ambeed
Warning

A111488-1G A111488-250MG
76982-742EA 245 USD
76982-742 76982-744
tert-Butyl 7-fluoro-6-nitro-3,4-dihydroisoquinoline-2(1H)-carboxylate 97%
tert-Butyl 7-fluoro-6-nitro-3,4-dihydroisoquinoline-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇FN₂O₄
MDL Number: MFCD22206990
CAS Number: 912846-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR