6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine 97%

Supplier: Ambeed
Warning

A111479-10G A111479-1G A111479-250MG A111479-25G A111479-5G
76995-236EA 297.06 USD
76995-236 76995-238 76995-240 76995-242 76995-244
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine 97%
6-Methyl-1H-pyrazolo[3,4-b]pyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₈N₄
MW: 148.165 g/mol
MDL Number: MFCD00456552
CAS Number: 79173-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR