3-(Bromomethyl)cyclobutanone ≥95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with calcium carbonate

A111249-100MG A111249-1G A111249-5G
76982-736EA 47.75 USD
76982-736 76982-738 77565-952
3-(Bromomethyl)cyclobutanone ≥95% stabilized
3-(Bromomethyl)cyclobutanone

Precaución: For research use only.

Formula: C₅H₇BrO
MDL Number: MFCD17170393
CAS Number: 463961-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR