Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate 95+%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-oxo-2,3-dihydrobenzo[d]oxazole-6-carboxylate

A111229-100MG A111229-1G A111229-250MG
76995-224EA 43.5 USD
76995-224 76995-226 76995-228
Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate 95+%
Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.16 g/mol
MDL Number: MFCD12827780
CAS Number: 72752-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR