4-(Hydroxymethyl)-1-methylpyrrolidin-2-one 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A111181-1G A111181-5G A111181-25G
76982-732EA 50 USD
76982-732 76982-734 77683-018
4-(Hydroxymethyl)-1-methylpyrrolidin-2-one 98%, Ambeed.Inc
4-(Hydroxymethyl)-1-methylpyrrolidin-2-one

Precaución: For research use only.

Formula: C₆H₁₁NO₂
MDL Number: MFCD09055222
CAS Number: 59887-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR