4-Cyanobenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Formylbenzonitrile

A111177-100G A111177-10G A111177-25G A111177-500G A111177-5G
76995-214EA 25 USD
76995-214 76995-216 76995-218 76995-220 76995-222
4-Cyanobenzaldehyde 97%
4-Cyanobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 262
MDL Number: MFCD00003376
CAS Number: 105-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR