4,7-Dibromo-2-phenylbenzo[d]thiazole 98%

Supplier: Ambeed
Warning

A111172-100MG A111172-1G A111172-250MG
77008-922EA 16.98 USD
77008-922 77008-924 77008-926
4,7-Dibromo-2-phenylbenzo[d]thiazole 98%
4,7-Dibromo-2-phenylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₁₃H₇Br₂NS
MW: 369.08 g/mol
MDL Number: MFCD27923419
CAS Number: 1588440-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR