4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Fluorophenyl)-6-isopropyl-2-[(N-methyl-N-methylsufonyl)amino]pyrimidine-5-yl-methanol
A110963-100G
A110963-10G
A110963-1G
A110963-25G
A110963-500G
A110963-5G
77025-764EA
175.05
USD
77025-764
77025-766
77025-768
77025-770
77025-772
77025-774
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol 97%
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methylsulfonylamino)pyrimidine-5-methanol
Precaución: for Research Use Only
Formula:
C₁₆H₂₀FN₃O₃S MW: 353.412 g/mol |
MDL Number:
MFCD08063917 CAS Number: 147118-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...