5-Bromo-2-(bromomethyl)pyrimidine 97%

Supplier: Ambeed
Danger

A110906-100MG A110906-250MG
77008-908EA 24 USD
77008-908 77008-910
5-Bromo-2-(bromomethyl)pyrimidine 97%
5-Bromo-2-(bromomethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MDL Number: MFCD16606298
CAS Number: 1193116-74-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR