2-(Methylthio)pyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A110776-1G A110776-250MG
77025-758EA 77 USD
77025-758 77025-760
2-(Methylthio)pyrimidine-5-carbonitrile 97%
2-(Methylthio)pyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MDL Number: MFCD11046326
CAS Number: 38275-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR