(S)-2-Amino-2-(4-fluorophenyl)ethanol 97%

Supplier: Ambeed
Warning

A110724-5G A110724-1G
76982-714EA 354 USD
76982-714 77570-608
(S)-2-Amino-2-(4-fluorophenyl)ethanol 97%
(S)-2-Amino-2-(4-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MDL Number: MFCD09253714
CAS Number: 224434-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR