4-Chloro-2-iodo-6-nitroaniline 95%

Supplier: Ambeed
Warning

A110708-1G A110708-250MG A110708-5G
76982-708EA 90 USD
76982-708 76982-710 76982-712
4-Chloro-2-iodo-6-nitroaniline 95%
4-Chloro-2-iodo-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄ClIN₂O₂
MDL Number: MFCD23115369
CAS Number: 123158-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR