3-(2,3-Dichlorophenyl)propionic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(2,3-Dichlorophenyl)propanoic acid
A110689-10G
A110689-1G
A110689-250MG
A110689-5G
76982-700EA
140.04
USD
76982-700
76982-702
76982-704
76982-706
3-(2,3-Dichlorophenyl)propionic acid 97%
3-(2,3-Dichlorophenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₉H₈Cl₂O₂ MW: 219.07 g/mol |
MDL Number:
MFCD06823953 CAS Number: 57915-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...