2-(4-Chlorobenzoyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)(pyridin-2-yl)methanone

A110480-100G A110480-10G A110480-1G A110480-25G A110480-5G
77025-734EA 172.93 USD
77025-734 77025-736 77025-738 77025-740 77025-742
2-(4-Chlorobenzoyl)pyridine 97%
2-(4-Chlorobenzoyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO
MW: 217.6500 g/mol
MDL Number: MFCD02930888
CAS Number: 6318-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR