5-Bromo-2,3-difluorobenzonitrile 95%

Supplier: Ambeed
Warning

A110406-100MG A110406-250MG A110406-1G
76982-696EA 25.47 USD
76982-696 76982-698 77565-944
5-Bromo-2,3-difluorobenzonitrile 95%
5-Bromo-2,3-difluorobenzonitrile

Precaución: For research use only.

Formula: C₇H₂BrF₂N
MW: 217.998 g/mol
MDL Number: MFCD15528910
CAS Number: 1105665-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR