3-Amino-6,7-difluoro-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 6,7-Difluoro-1H-indazol-3-amine

A110397-1G A110397-250MG A110397-5G
77025-728EA 59.42 USD
77025-728 77025-730 77025-732
3-Amino-6,7-difluoro-1H-indazole 95%
3-Amino-6,7-difluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅F₂N₃
MW: 169.1300 g/mol
MDL Number: MFCD13191632
CAS Number: 706805-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR