3-Methoxyoxan-4-one 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methoxy-tetrahydropyran-4-one, 3-Methoxydihydro-2H-pyran-4(3H)-one
A110381-1G
A110381-250MG
A110381-25G
A110381-5G
76995-172EA
61.54
USD
76995-172
76995-174
76995-176
76995-178
3-Methoxyoxan-4-one 95%
3-Methoxyoxan-4-one
Precaución: for Research Use Only
Formula:
C₆H₁₀O₃ MW: 130.142 g/mol |
MDL Number:
MFCD09909903 CAS Number: 624734-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...