6-(Difluoromethoxy)pyridin-2-amine 98%

Supplier: Ambeed
Warning

A110334-100MG A110334-1G
76982-694EA 112.46 USD
76982-694 77683-013
6-(Difluoromethoxy)pyridin-2-amine 98%
6-(Difluoromethoxy)pyridin-2-amine

Precaución: For research use only.

Formula: C₆H₆F₂N₂O
MW: 160.12 g/mol
MDL Number: MFCD13185863
CAS Number: 1131007-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR