6-Methoxy-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A110283-100MG A110283-250MG A110283-1G A110283-5G
77025-724EA 32 USD
77025-724 77025-726 77599-584 77599-586
6-Methoxy-1H-pyrrolo[2,3-b]pyridine 97%
6-Methoxy-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD06659665
CAS Number: 896722-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR