3-Chloro-7,8-dihydroquinolin-5(6H)-one 95%

Supplier: Ambeed
Warning

A110155-100MG A110155-250MG
77025-720EA 53.05 USD
77025-720 77025-722
3-Chloro-7,8-dihydroquinolin-5(6H)-one 95%
3-Chloro-7,8-dihydroquinolin-5(6H)-one

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.619 g/mol
MDL Number: MFCD10696330
CAS Number: 127724-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR