2,3,4,5-Tetrahydrobenzo[f][1,4]oxazepine 97%

Supplier: Ambeed
Warning

A110150-100MG A110150-250MG A110150-1G
77025-716EA 145.35 USD
77025-716 77025-718 77666-604
2,3,4,5-Tetrahydrobenzo[f][1,4]oxazepine 97%
2,3,4,5-Tetrahydrobenzo[f][1,4]oxazepine

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD08669765
CAS Number: 17775-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR