2-Aminopyrimidin-5-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-hydroxypyrimidine

A110115-100MG A110115-250MG
76982-682EA 79.57 USD
76982-682 77683-012
2-Aminopyrimidin-5-ol 97%
2-Aminopyrimidin-5-ol

Precaución: For research use only.

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD08062411
CAS Number: 143489-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR