(S)-(+)-2-Aminobutyric acid 97%

Supplier: Ambeed
Warning

A110043-1000G A110043-100G A110043-10G A110043-25G A110043-500G
76995-156EA 56 USD
76995-156 76995-158 76995-160 76995-162 76995-164
(S)-(+)-2-Aminobutyric acid 97%
(S)-(+)-2-Aminobutyric acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD00064415
CAS Number: 1492-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR