4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine 95%
Supplier: Ambeed
|
Warning
|
A110040-100MG
A110040-10G
A110040-1G
A110040-250MG
A110040-5G
77025-706EA
49.87
USD
77025-706
77025-708
77025-710
77025-712
77025-714
4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine 95%
4-(3-Bromoimidazo[1,2-b]pyridazin-6-yl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrN₄O MW: 283.1300 g/mol |
MDL Number:
MFCD11520886 CAS Number: 1012343-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...