6-Chloro-2-isopropyl-1H-benzo[d]imidazole 98%

Supplier: Ambeed
Warning

A110007-1G A110007-250MG
77008-864EA 113.52 USD
77008-864 77008-866
6-Chloro-2-isopropyl-1H-benzo[d]imidazole 98%
6-Chloro-2-isopropyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
MW: 194.66 g/mol
MDL Number: MFCD18800924
CAS Number: 4886-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR