2-(tert-Butyl)-4-nitrophenol 95%

Supplier: Ambeed
Warning

A109901-100MG A109901-250MG
76982-670EA 83.82 USD
76982-670 76982-672
2-(tert-Butyl)-4-nitrophenol 95%
2-(tert-Butyl)-4-nitrophenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.21 g/mol
MDL Number: MFCD01463425
CAS Number: 6683-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR