(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2,2'-((5-Amino-1-carboxypentyl)azanediyl)diacetic acid, Nα,Nα-Bis(carboxymethyl)-L-lysine
A109900-1G
A109900-250MG
A109900-10G
A109900-100G
A109900-5G
76982-666EA
42.44
USD
76982-666
76982-668
77565-932
77683-004
77683-005
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid 95%, Ambeed.Inc
(5S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid
Precaución: For research use only.
Formula:
C₁₀H₁₈N₂O₆ MW: 262.26 g/mol |
MDL Number:
MFCD00237437 CAS Number: 113231-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...