2-Bromo-5-(trifluoromethyl)pyrimidine 95%

Supplier: Ambeed
Warning

A109703-100MG A109703-250MG A109703-1G
76982-648EA 70.02 USD
76982-648 76982-650 77549-116
2-Bromo-5-(trifluoromethyl)pyrimidine 95%
2-Bromo-5-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂
MW: 226.982 g/mol
MDL Number: MFCD14702304
CAS Number: 69034-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR