2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine 95%

Supplier: Ambeed
Warning

A109692-1G A109692-250MG A109692-100MG
77025-702EA 238 USD
77025-702 77025-704 77666-600
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine 95%
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₉H₁₅N₅
MDL Number: MFCD13193411
CAS Number: 943757-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR