Methyl 5-fluoro-3-methylpicolinate 98%

Supplier: Ambeed
Warning

A109616-100MG A109616-250MG
77008-858EA 66.84 USD
77008-858 77008-860
Methyl 5-fluoro-3-methylpicolinate 98%
Methyl 5-fluoro-3-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
MDL Number: MFCD22560016
CAS Number: 1346148-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR