7-Bromoisatin 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione

A109571-100G A109571-10G A109571-1G A109571-25G A109571-500G A109571-5G
76995-132EA 210.06 USD
76995-132 76995-134 76995-136 76995-138 76995-140 76995-142
7-Bromoisatin 95%
7-Bromoisatin

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
MDL Number: MFCD00774354
CAS Number: 20780-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR