1-(4-Chloro-2-fluoro-6-hydroxyphenyl)ethanone 95%

Supplier: Ambeed
Warning

A109532-100MG A109532-1G A109532-250MG
77025-680EA 53.05 USD
77025-680 77025-682 77025-684
1-(4-Chloro-2-fluoro-6-hydroxyphenyl)ethanone 95%
1-(4-Chloro-2-fluoro-6-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
MDL Number: MFCD13152285
CAS Number: 1110663-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR