[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one 95%

Supplier: Ambeed
Warning

A109492-100MG A109492-250MG
77008-850EA 45 USD
77008-850 77008-852
[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one 95%
[1,2,4]Triazolo[4,3-a]pyrimidin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₄N₄O
MDL Number: MFCD13193348
CAS Number: 63206-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR