5-Bromo-2-chloronicotinaldehyde 98%

Supplier: Ambeed
Warning

A109483-100MG A109483-1G A109483-250MG A109483-5G
77008-844EA 11 USD
77008-844 77008-846 77008-848 77570-538
5-Bromo-2-chloronicotinaldehyde 98%
5-Bromo-2-chloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
MDL Number: MFCD08689713
CAS Number: 228251-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR