L-(-)-Tryptophanol 97%

Supplier: Ambeed
Warning

Synonyms: L-Tryptophanol, (S)-(-)-2-Amino-3-(3-indolyl)propanol, (S)-2-Amino-3-(1H-indol-3-yl)propan-1-ol

A109445-1G A109445-250MG A109445-5G
76995-112EA 44.56 USD
76995-112 76995-114 76995-116
L-(-)-Tryptophanol 97%
L-(-)-Tryptophanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
MDL Number: MFCD00037970
CAS Number: 2899-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR