2,5,6-Trichloro-4-methylnicotinonitrile 98%

Supplier: Ambeed
Warning

A109435-100MG A109435-1G A109435-250MG
77025-668EA 8.49 USD
77025-668 77025-670 77025-672
2,5,6-Trichloro-4-methylnicotinonitrile 98%
2,5,6-Trichloro-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₃N₂
MW: 221.47 g/mol
MDL Number: MFCD09754021
CAS Number: 63195-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR