4-Cyano-2,3-difluorobenzoic acid 95%

Supplier: Ambeed
Danger

A109353-100MG A109353-1G A109353-5G
76982-642EA 22 USD
76982-642 76982-644 76982-646
4-Cyano-2,3-difluorobenzoic acid 95%
4-Cyano-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃F₂NO₂
MDL Number: MFCD20640528
CAS Number: 1033997-04-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR