3-Bromo-5-fluoro-4-methylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-bromo-6-fluorotoluene, 3-Bromo-5-fluoro-p-toluidine

A109351-100MG A109351-250MG A109351-5G A109351-1G
76982-636EA 42.44 USD
76982-636 76982-638 76982-640 77570-536
3-Bromo-5-fluoro-4-methylaniline 98%
3-Bromo-5-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.04 g/mol
MDL Number: MFCD12025433
CAS Number: 207110-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR