1-(5-Bromo-7-azaindole)ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone

A109245-1G A109245-250MG
76982-632EA 26 USD
76982-632 77889-758
1-(5-Bromo-7-azaindole)ethanone 98%
1-(5-Bromo-7-azaindole)ethanone

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MDL Number: MFCD12403936
CAS Number: 866545-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR