Z-Hyp-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-4-hydroxy-L-proline, Z-L-4-Hydroxyproline, trans-N-Cbz-4-hydroxy-L-proline, (2S,4R)-1-((Benzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, N-Cbz-Hydroxy-L-Proline

A109231-100G A109231-10G A109231-25G A109231-500G A109231-5G
76995-102EA 54.11 USD
76995-102 76995-104 76995-106 76995-108 76995-110
Z-Hyp-OH 95%
Z-Hyp-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₅
MW: 265.3 g/mol
MDL Number: MFCD00037329
CAS Number: 13504-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR