2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole 97%
Supplier: Ambeed
|
Warning
|
A109228-100G
A109228-250MG
A109228-500G
A109228-25G
A109228-5G
77025-658EA
77
USD
77025-658
77025-660
77670-546
77880-886
77880-887
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole 97%
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole
Precaución: for Research Use Only
Formula:
C₁₆H₁₄F₃N₃OS |
MDL Number:
MFCD00834357 CAS Number: 103577-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...