Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-bromopyrazolo[1,5-a]pyridine-3-carboxylate
A109203-100MG
A109203-1G
A109203-250MG
A109203-5G
A109203-25G
77008-826EA
38.2
USD
77008-826
77008-828
77008-830
77008-832
77549-102
Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate 98%
Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₉BrN₂O₂ MW: 269.1 g/mol |
MDL Number:
MFCD06739182 CAS Number: 885276-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...