5-Iodo-2-methoxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A109039-1G
A109039-250MG
A109039-5G
76982-624EA
103.97
USD
76982-624
76982-626
76982-628
5-Iodo-2-methoxybenzoic acid 95%
5-Iodo-2-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ MW: 278.04 g/mol |
MDL Number:
MFCD02683859 CAS Number: 2786-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...